Få mere at vide
Masterflex™ Polycarbonate Manifolds
Artikelnummer. 11785239
Antal havne:
2
3
4
5
Inkluderer:
180° Rotation
360° Rotation
Pakningsstørrelse:
Hver
Artikelnummer. 11785239
Brand: Masterflex™ 0646485
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulate flow without leaking with these manifolds with luer locks. Cole Parmer™ Polycarbonate Manifolds feature polyurethane stems, HDPE handles and female ports and luer locks.
  • Purchase manifolds individually or in convenient packs of ten
  • This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Tekniske data
Tilslutningstype Luer Fitting
Oversigt 180°
Inkluderer 180° Rotation
Materiale Polycarbonate
Antal havne 4