missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Polybutenes, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Polybutenes |
| CAS | 9003-29-6 |
| Molekylær formel | [C4H8]x . [C4H8]y |
| Mængde | 10 mL |
| Synonym | Amoco 15H, B 912, Beaulon P 5040, Butene polymer, C 9900, Chevron 12, Chevron 16, Chevron 18, Chevron 6, Cosden OSH, DP 8510M, Duraflex 8510, Dynapak H 2000, EIM 8, Glissopal 10, Glissopal 3251, Glissopal 91051, Glissopal 92485, Glissopal 93026, Glissopal 93039, Glissopal CE 5203, H 15, H 25, HE 175, HE 1975, HE 300, HE 375, HR 2300, HRPB 1300, HT 300, HV 100, HV 100F(SB), HV 15, HV 15E, HV 1900F, HV 300F, HV 300R, HV 30SH, HV 35, Hyvis 10, Hyvis 3, Hyvis 7000/45, Idemitsu Polybutene 10H-A, Idemitsu Polybutene 15R, Idemitsu, Polybutene 3000, Idemitsu Polybutene 300H, Idemitsu Polybutene 5H, Idemitsu Polybutene OH, Indopol 1900, Indopol 500, Indopol H 15, Indopol H 1800, Indopol H 18000, Indopol H 200, Indopol H 25, Indopol H 40, Indopol H 7, Indopol L 10, Indopol L 4, L 14, L 14 (polymer), LP 15, LP 15 (polyolefin), LV 100, LV 50, LZV 100, NA (solvent), NA Solvent NAS 5H, NAS 5H, Napvis 2000, Napvis 3, Napvis 600, Napvis D 30, Napvis DE 10, Neosealer MS-N 12, Newglide M, Nissan Polybutene 015N, Nisseki HV, Oktol 600, Oloa 9350, Oronite 6, PO 300H, Parapol 450, Parapol 700, Petrofin 100, Poly SH 1900, Poly SH 2500, Poly-n-butene, Polybuden 300H, Polybutene 06N, Polybutene 0H, Polybutene 100H, Polybutene 10N, Polybutene 15R, Polybutene 2000, Polybutene 300H, Polybutene 5SH, Polybutene HT-A, Polybutene HV 100, Polybutene HV 35, Polybutene LV 50, Polybutene ON, Polybutene PB 5H, Polybutene SH 015, Polybutylene, Polyvis, Polyvis 015N, Polyvis 025SH, Polyvis 06N, Polyvis 06SH, Polyvis 0N, Polyvis 10N, Polyvis 150SH, Polyvis 2000CH, Polyvis 200SH, Polyvis 300H, Polyvis 3SH, Polyvis 5N, Polyvis NC 7000, Polyvis ON, Polyvis OO, RHV 300, ResinKit 22, SV 7000, Slovis 500, Slovis D 400, Slovis W 800, Soltex 124, Tafmer BL 027, Tafmer M 2481, Ultravis 120, VH 100, Vestowax B 912, Witron 0200, Witron 131 |
| InChI formel | InChI=1S/C4H10/c1-3-4-2/h3-4H2,1-2H3 |
| SMIL | CCC(C)(C)CC(CC)C |
| Molekylvægt (g/mol) | 112.21 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?