missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Poly(trifluorovinyl chloride), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Poly(trifluorovinyl chloride) |
| CAS | 9002-83-9 |
| Molekylær formel | [CF2CF(Cl)]n |
| Mængde | 1 g |
| Synonym | Aclar RX 160, CTFE oil 27, CTFE oil 56, Chlorotrifluoroethene homopolymer, Chlorotrifluoroethylene homopolymer, Chlorotrifluoroethylene polymer, Clarus P 4000TR, DF 0025C1, DF 1, Daifloil 10, Daifloil 100, Daifloil 20, Daifloil 3, Daifloil 50, Daiflon CTFE, Daiflon CTFE-D 55P, Daiflon D 45S, Daiflon M 300, Daiflon M 300P, EL Sealer 4810N-R, EL Sealer No. 4810N-R, Ekafluvin, F 3, F 3 (vinyl polymer), FL-G 100, FL-G 330, FL-G 35, FL-G 5, FLuorolube LG 160, Flonlube FL 500, Flonlube FL 800, Fluorolon 3, Fluorolube 2000, Fluorolube 300/140, Fluorolube FS 5, Fluorolube GR 470, Fluorolube S 30, Fluoroplast 3, Fluoroplast3P, Fluoroplast F 3, Fluorothene, Flurolube FS 5, Ftorlon 3, Ftoroplast 3, Ftoroplast 3P, Fulusin T, HBF 430, HFM-F 8800, Halar 200, HaloVac 60, Halocarbon 95, Halon, Hostaflon C, Kel S, Kel-F, Kel-F 10, Kel-F 200, Kel-F 3, Kel-F 6050, Kel-F 6061, Kel-F700, Kel-F 81, Kel-F 90, Kel-F 95/5, Kel-F oil, M 300, M 300P, M 400H, M 400P, MLO 81-125, MLO 83-322, Monochlorotrifluoroethylene polymer, Neoflen CTFE-M 400, Neoflon CTFE, Neoflon CTFE-M 300, Neoflon CTFE-M 300PL, Neoflon CTFE-M 400, Neoflon CTFE-M 400H, Neoflon DF 0025, Neoflon DF 0025C, Neoflon DF 0050C1, Neoflon M 300, Neoflon M 300P, Neoflon M 300PL, Neoflon M 400H, Neoflon PCTFE, Neoflon PCTFE-DF0050C1, Neoflon PCTFE-M 300, Neosorb ND, P 2000TR, PCTFE, Plaskon CTFE, Poly(1-chloro-1,2,2-trifluoroethylene), Poly(chlorotrifluoroethene), Poly(chlorotrifluoroethylene), Poly(ethylene trifluoride chloride), Poly(monochlorotrifluoroethylene), Poly(trifluorochloroethylene), Poly(trifluoroethylenechloride), Poly(trifluoromonochloroethylene), Poly(trifluorovinyl chloride), Poly(vinyl trifluorochloride), Safetol 3.1, TBM 240, TL 340, Teflex, Teflex(fluoropolymer), Teflon PCTFE, Trifluorochloroethylene homopolymer, Trifluorochloroethylene polymer, Voltalef, Voltalef 10S, Voltalef 1S, Voltalef 202, Voltalef 300LD, Voltalef 300UF, Voltalef 302, Voltalef UF 300 |
| InChI formel | InChI=1S/C2ClF3/c3-1(4)2(5)6 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?