missing translation for 'onlineSavingsMsg'
Få mere at vide

Poly(trifluorovinyl chloride), TRC

Artikelnummer. 30418219
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418219 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30418219 Leverandør Toronto Research Chemicals Leverandørnr. TRCP68939010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Poly(trifluorovinyl chloride)
CAS 9002-83-9
Molekylær formel [CF2CF(Cl)]n
Mængde 10 g
Synonym Aclar RX 160, CTFE oil 27, CTFE oil 56, Chlorotrifluoroethene homopolymer, Chlorotrifluoroethylene homopolymer, Chlorotrifluoroethylene polymer, Clarus P 4000TR, DF 0025C1, DF 1, Daifloil 10, Daifloil 100, Daifloil 20, Daifloil 3, Daifloil 50, Daiflon CTFE, Daiflon CTFE-D 55P, Daiflon D 45S, Daiflon M 300, Daiflon M 300P, EL Sealer 4810N-R, EL Sealer No. 4810N-R, Ekafluvin, F 3, F 3 (vinyl polymer), FL-G 100, FL-G 330, FL-G 35, FL-G 5, FLuorolube LG 160, Flonlube FL 500, Flonlube FL 800, Fluorolon 3, Fluorolube 2000, Fluorolube 300/140, Fluorolube FS 5, Fluorolube GR 470, Fluorolube S 30, Fluoroplast 3, Fluoroplast3P, Fluoroplast F 3, Fluorothene, Flurolube FS 5, Ftorlon 3, Ftoroplast 3, Ftoroplast 3P, Fulusin T, HBF 430, HFM-F 8800, Halar 200, HaloVac 60, Halocarbon 95, Halon, Hostaflon C, Kel S, Kel-F, Kel-F 10, Kel-F 200, Kel-F 3, Kel-F 6050, Kel-F 6061, Kel-F700, Kel-F 81, Kel-F 90, Kel-F 95/5, Kel-F oil, M 300, M 300P, M 400H, M 400P, MLO 81-125, MLO 83-322, Monochlorotrifluoroethylene polymer, Neoflen CTFE-M 400, Neoflon CTFE, Neoflon CTFE-M 300, Neoflon CTFE-M 300PL, Neoflon CTFE-M 400, Neoflon CTFE-M 400H, Neoflon DF 0025, Neoflon DF 0025C, Neoflon DF 0050C1, Neoflon M 300, Neoflon M 300P, Neoflon M 300PL, Neoflon M 400H, Neoflon PCTFE, Neoflon PCTFE-DF0050C1, Neoflon PCTFE-M 300, Neosorb ND, P 2000TR, PCTFE, Plaskon CTFE, Poly(1-chloro-1,2,2-trifluoroethylene), Poly(chlorotrifluoroethene), Poly(chlorotrifluoroethylene), Poly(ethylene trifluoride chloride), Poly(monochlorotrifluoroethylene), Poly(trifluorochloroethylene), Poly(trifluoroethylenechloride), Poly(trifluoromonochloroethylene), Poly(trifluorovinyl chloride), Poly(vinyl trifluorochloride), Safetol 3.1, TBM 240, TL 340, Teflex, Teflex(fluoropolymer), Teflon PCTFE, Trifluorochloroethylene homopolymer, Trifluorochloroethylene polymer, Voltalef, Voltalef 10S, Voltalef 1S, Voltalef 202, Voltalef 300LD, Voltalef 300UF, Voltalef 302, Voltalef UF 300
InChI formel InChI=1S/C2ClF3/c3-1(4)2(5)6
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.