Learn More
Schneider Gerd™ Poly Spoon
Use this spoon for a variety of applications. Schneider Gerd™ Poly Spoons are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 2994
Additional Details : Weight : 0.10000kg
Product Code. 10682835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Spoons collect and transport dry substances or liquids
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
Order Info
LengthMetric
Specifications
Specifications
1 Pk.
Poly Spoon
190 mm