missing translation for 'onlineSavingsMsg'
Learn More

Poly(methylhydrosiloxane), trimethylsilyl terminated, Thermo Scientific Chemicals

Catalog Number p-7077279
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
63148-57-2
(CH4OSi)n
MFCD00084478
QNWOFLWXQGHSRH-UHFFFAOYSA-N
C[SiH](O[Si](C)(C)C)O[Si](C)(C)C
This item is not returnable. View return policy
63148-57-2
(CH4OSi)n
MFCD00084478
QNWOFLWXQGHSRH-UHFFFAOYSA-N
C[SiH](O[Si](C)(C)C)O[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0060g/mL
Flash Point 204°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3H9OSi-(CH4OSi)n-CH3H9Si
Quantity 25 mL
Specific Gravity 1.006
Chemical Name or Material Poly(methylhydrosiloxane)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'