Få mere at vide
Poly(hexamethylenebicyanoguanide-hexamethylenediamine) Hydrochloride(Technical Grade), TRC
Artikelnummer. 30132391
Skift visning
Mængde:
1 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132391 50 g
30005599 25 g
30008873 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132391 Leverandør Toronto Research Chemicals Leverandørnr. TRCP68920050G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27083-27-8
Molekylær formel (C10H18N8 . C6H16N2)x . HCl
Synonym N,N' ' '-1,6-Hexanediylbis[N'-cyanoguanidine, polymer with 1,6-Hexanediamine Hydrochloride
IUPAC navn 1-cyano-3-[6-[(N-cyanocarbamimidoyl)amino]hexyl]guanidine;hydrochloride
SMIL N=C(NCCCCCCNC(NC#N)=N)NC#N.Cl
Tekniske data
Kemisk navn eller materiale Poly(hexamethylenebicyanoguanide-hexamethylenediamine) Hydrochloride
Mængde 50 g
InChI formel InChI=1S/C10H18N8.ClH/c11-7-17-9(13)15-5-3-1-2-4-6-16-10(14)18-8-12;/h1-6H2,(H3,13,15,17)(H3,14,16,18);1H
Formel vægt 286.1421