Learn More
Poly(ethylene oxide), approx. M.W. 600,000
Product Code. 10534121 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10534121 250 g Glass bottle 250g
10751261 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10534121 Supplier Thermo Scientific Supplier No. 178612500
 Restricted Item
Forventet forsendelse: 20-11-2026
Add to basket
Edge
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
Specifications
Color White
Physical Form Granular Powder
Quantity 250 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass bottle
Linear Formula (-CH2CH2O-)n
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Chemical Name or Material Poly(ethylene oxide), Approximate M.W. 600.000