Få mere at vide
Poly(ethylene glycol) Monooleate (Technical Grade ~70%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Polyoxyl Oleate |
| CAS | 9004-96-0 |
| Molekylær formel | (C2H4O)nC18H34O2 |
| Mængde | 25 mL |
| Synonym | Adeka Estol OEG 204, Agnique PEG 400MO, Akyporox O 50, Alkamuls 400MO, Alkamuls A, Alkamuls A 2, Alkasurf O 14, Alkasurf O 75-9, AtlasG 2140, Blaunon EN 1507, CRL 1337, Cemulsol 1050, Chemax E 400MO, Chemester 300OC, ChimiponOCD, Cithrol 2MO, Cithrol PO, Crodet O 100, Dyapol G, Emalex 218, Emanon 4110, Emanon 4115, Emcol H 2A, Emcol H 31A, Emerest 2624, Emerest 2646, Emerest 2660, Empilan BP 100, Empilan BQ100, Emulan A, Emulphor 24, Estax 38 S.F, Ethofat O, Ethox MO 14, Ethoxylated oleic acid, Ethylan A 2, Ethylene oxide-oleic acid adduct, Extrex P 60, G 2142, G2143, G 2144, G 5507, H 2A, H 3549A, Hamadol OD 600, ITRI-PH 02, Interwet 33, Ionet MO 200, Ionet MO 400, Ionet MO 600, Ionet PO 600, KEOL15, Kessco PEG 1000MO, Kessco PEG 400MO, Konion MO 14, Koremul OA 9, Lannagol LF, Ligalub PEG 400MO, Lionon MO 60, Lipal 300W, Lipal30W, Lipal 400OL, Lipo-Peg 4O, MO 600, MYO 2, Macrogol oleate, Macrogol oleate 600, Mapeg 200, Marlosol OL 15, Marlosol OL 7, Mergital OA 6, Monooleate ester of polyethylene glycol, Newcol 170, Nikkol MYO 10, Nissan Nonion O 2, Noigen ES 120, 200, Nopalcol 1O, Oleic acid poly(oxyethylene) ester, Oleopal 1, Oleox 3, PEG 1000MO, PEG 200 oleate, PEG 200MO, PEG 300 monooleate, PEG-400 monoester with oleic acid, Pegnol O 14, Pegosperse 400MO, Peretex E 451D, Pionin D 2507A, Plasthall CPH 41N, Poly(ethylene glycol) oleic acidester, Poly(ethylene oxide) oleate, Poly(oxyethylene) monooleate, Poly(oxyethylene) oleate, Poly(oxyethylene)oleic acid ester, Polyethylene glycol monooleate, Polyethylene glycol oleate, Polyethylene oxide monooleate, Polyglycol monooleate, Polyglycol oleate, Polynon O 44, Polyoxyethylene ether oleate, Polyoxyethylene monooleic acid ester, Prodhyphore B, Radiasurf 7402, Rikemal OE 809, Rokacet O 7, S 1006, S 1132, Serdox NOG 264, Slovasol A, Standapol 2660, Surfonic E 400MO, Teric OF 6, Texafor M 6, Tilol 165, Trydet OA 7, Unichema A, Unisol 4-0, Wardol X, Witco 31, Witconol H 31A, Witconol H 35A, X539R |
| InChI formel | InChI=1S/C20H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(22)23-19-18-21/h9-10,21H,2-8,11-19H2,1H3/b10-9- |
| SMIL | O=C(OCCO)CCCCCCC/C=C/CCCCCCCC |
| Molekylvægt (g/mol) | 326.51 |
| Formel vægt | 326.2821 |