missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000, Thermo Scientific Chemicals

Catalog Number p-185701
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
9003-01-4
(C3H4O2)n
72.06
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
6581
CHEBI:18308
OC(=O)C(-*)C-*
This item is not returnable. View return policy
9003-01-4
(C3H4O2)n
72.06
MFCD00084394
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
6581
CHEBI:18308
OC(=O)C(-*)C-*

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Linear Formula [-CH2CH(CO2H)-]n
Solubility Information Solubility in water: soluble.
Quantity 1 g
IUPAC Name prop-2-enoic acid
Formula Weight 1.080.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 10% max.
Packaging Glass bottle
Chemical Name or Material Poly(acrylic acid)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'