missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ POLR2L Recombinant Protein Antigen
Recombinant Protein
Brand: Novus Biologicals™ NBP3-17418PEP
Additional Details : Weight : 0.00970kg
Specifications
Antibody Competition | |
5441 | |
100 μg | |
DNA-directed RNA polymerase III subunit L, DNA-directed RNA polymerases I, II, and III subunit RPABC5, EC 2.7.7.6, hRPB7.6, hsRPB10b, polymerase (RNA) II (DNA directed) polypeptide L (7.6kD), polymerase (RNA) II (DNA directed) polypeptide L, 7.6kDa, RBP10, RNA polymerase II 7.6 kDa subunit, RPABC5, RPB10, RPB10 homolog, RPB10beta, RPB7.6RNA polymerases I, II, and III subunit ABC5 | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
POLR2L |
PBS and 1M Urea, pH 7.4. | |
M.W. Theoretical: 21 kDa | |
Store at -20°C. Avoid freeze-thaw cycles. | |
POLR2L | |
>80% by SDS-PAGE and Coomassie blue staining |