Få mere at vide
Polixetonium Chloride, TRC
Artikelnummer. 30133834
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133834 10 g
30135015 50 g
17921975 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133834 Leverandør Toronto Research Chemicals Leverandørnr. TRCP68809310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 31512-74-0
Molekylær formel (C10H24Cl2N2O)n
Molekylvægt (g/mol) 289.28
Synonym Poly[oxy-1,2-ethanediyl(dimethyliminio)-1,2-ethanediyl(dimethyliminio)-1,2-ethanediyl chloride (1:2)] (ACI), Poly[oxy-1,2-ethanediyl(dimethyliminio)-1,2-ethanediyl(dimethyliminio)-1,2-ethanediyl dichloride] (9CI), Poly[oxyethylene(dimethyliminio)ethylene(dimethyliminio)ethylene dichloride] (8CI), Armoblen NPX, BL 2142, Bualta, Bubond 60, Bulab 6002, Busan 1507, Busan 77, KA 1700, MBC 115, Polixetonium chloride, Poly[oxyethylene(dimethylamino)ethylene(dimethylamino)ethylene dichloride], Poly[oxyethylene(dimethylimino)ethylene(dimethylimino)ethylene dichloride], Polyquaternium 42, TB 66, WSCP
SMIL [Cl-].[Cl-].CCC[N+](C)(C)CC[N+](C)(C)CCOC
Tekniske data
Kemisk navn eller materiale Polixetonium Chloride
Mængde 10 g
InChI formel InChI=1S/C12H30N2O.2ClH/c1-7-8-13(2,3)9-10-14(4,5)11-12-15-6;;/h7-12H2,1-6H3;2*1H/q+2;;/p-2
Formel vægt 258.1266