Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter for Transmitted Light Stage (122 150)
Turnable Polarization Filter attachment. Analyzer needs to be ordered separately.
Brand: SCHOTT AG Lighting and Imaging™ 158500
Additional Details : Weight : 0.10000kg
Description
- Provides polarized light by a polarizing and analyzing filter within the beam path.
- Can be very efficiently used in Stereo Microscopy.
- Helps to contrast some inner structure of material or to identify metal particles within others.
Specifications
Polarization Filter | |
Stereo Microscopy Illumination | |
SCHOTT Transmitted Light Stage 122 150 |