Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter for Transmitted Light Stage (122 150)
Turnable Polarization Filter attachment. Analyzer needs to be ordered separately.
Brand:  SCHOTT AG Lighting and Imaging™ 158500
Additional Details : Weight : 0.10000kg
Product Code. 10269411
  • 1280.00 DKK / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket
Description
Description
  • Provides polarized light by a polarizing and analyzing filter within the beam path.
  • Can be very efficiently used in Stereo Microscopy.
  • Helps to contrast some inner structure of material or to identify metal particles within others.
Specifications
Specifications
Polarization Filter
Stereo Microscopy Illumination
SCHOTT Transmitted Light Stage 122 150