missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Polaprezinc, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Polaprezinc |
| CAS | 107667-60-7 |
| Molekylær formel | C9 H12 N4 O3 Zn |
| Mængde | 100 mg |
| Synonym | Zinc','[β-alanyl-κN-L-histidinato(2-)-κN,κO]- (9CI), L-Histidine','N-β-alanyl-','zinc complex (ZCI), Zinc','[N-β-alanyl-L-histidinato(2-)-N,NN,Oα]- (ZCI), Polaprezinc, Promac, Promac (antiulcer agent), Z 103, Zinc L-carnosine, β-Alanyl-L-histidinato zinc, [β-Alanyl-κN-L-histidinato(2-)-κN,κO]zinc |
| InChI formel | InChI=1S/C9H14N4O3.Zn/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6;/h4-5,7H,1-3,10H2,(H3,11,12,13,14,15,16);/q;+2/p-2/t7-;/m0./s1 |
| SMIL | NCCC(=O)N1[Zn]OC(=O)[C@@H]1Cc2cnc[nH]2 |
| IUPAC navn | (4S)-3-(3-aminopropanoyl)-4-(1H-imidazol-5-ylmethyl)-1-oxa-3-aza-2λ^{2}-zincacyclopentan-5-one |
| Molekylvægt (g/mol) | 289.63 |
| Formel vægt | 288.0201 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?