Learn More
Sampling Systems™ Pointed Scissors
Product Code. 17902102 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
310 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
17902102 310 mm Each
1 options
Product Code. 17902102 Supplier Sampling Systems™ Supplier No. A364300
  • ONLINE EXCLUSIVE PRICE
    156.00 DKK
    150.35 DKK / Pakke med 1
    Spare 5.65 DKK
    3% Slukket
Forventet forsendelse: 14-01-2027
High quality pointed scissors made from surgical grade 420 stainless steel.
  • Ideal for cutting and manipulating samples
Specifications
Blade Material Stainless Steel
Handle Type Loop
Product Type Surgical Scissors
Blade Style Straight
Color-coded No
Grade Hospital
Handle Material Stainless Steel
Length (English) 12.2 in.
Length (Metric) 310 mm
Length (Metric) Blade 190 mm
Material Stainless Steel
Size Long
Autoclavable Autoclavable
Color Silver
Cut Type Shear
Sterility Non-sterile
Tip Style Pointed