missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ PODN Polyclonal Antibody

Beskrivelse
Antibody detects endogenous levels of total PODN.
Non-collagenous proteins of the small leucine-rich repeat (SLR) protein family are considered important components of the extracellular matrix. The extracellular matrix plays an integral role in the pivotal processes of development, tissue repair and metastasis by regulating cell proliferation, differentiation, adhesion and migration. Podocan, also known as PCAN, SLRR5A or PODN, is a 613 amino acid secreted protein belonging to the small leucine-rich proteoglycan (SLRP) family and the class V subfamily. Expressed in kidney, heart, liver, pancreas and vascular smooth muscle cells, Podocan exists as three alternatively spliced isoforms containing twenty LRR (leucine-rich repeats), a unique N-terminal cysteine-rich cluster pattern and a highly acidic C-terminal domain. Podocan is considered a glycoprotein containing N-linked oligosaccharides that may be involved in growth regulation in cardiovascular tissues. Encoded by a gene located on human chromosome 1, Podocan interacts type I collagen (COL1).
Tekniske data
Tekniske data
| Antigen | PODN |
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Podn |
| Genadgangsnr. | Q7Z5L7 |
| Gene Alias | 9430070G18; Pcan; Podn; Podocan; podocan proteoglycan; PRO332; SLRR5A; UNQ293; UNQ293/PRO332 |
| Gensymboler | Podn |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?