Learn More
BRAND™ Rack for Sedimentation Cones
Holds 2 Imhoff sedimentation cones made of plastic. Compact design and easy to carry, even with cones filled.
870.00 DKK - 985.00 DKK
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
10048850
|
BRAND™
388050 |
300 x 130 x 315mm |
870.00 DKK
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
10606901
|
BRAND™
388060 |
300 x 130 x 400mm |
985.00 DKK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Holds 2 Imhoff sedimentation cones
- Polymethyl methacrylate (PMMA)/Polypropylene (PP) construction
Specifications
Imhoff Cone Rack | |
PMMA, Polypropylene |