missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PML Protein Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-98823FR
Additional Details : Weight : 0.00970kg
Description
PML Protein Polyclonal specifically detects PML Protein in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PML Protein | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 680 | |
MYLPP8675, promyelocytic leukemia, Promyelocytic leukemia protein, RING finger protein 71, RNF71probable transcription factor PML, TRIM19promyelocytic leukemia, inducer of, tripartite motif protein TRIM19, Tripartite motif-containing protein 19 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human PML Protein. | |
0.1 mL | |
Cancer, Zinc Finger | |
5371 | |
Store at 4°C in the dark. | |
IgG |