missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies Pluripotin
Beskrivelse
- Pluripotin inhibits both Ras GTPase-activating protein (RasGAP, Kd = 98 nM) and extracellular signal-regulated kinase 1 (ERK1, Kd = 212 nM; Chen et al.) Inhibition of RasGAP increases Ras signaling via the PI3-kinase pathway which promotes self-renewal, whereas inhibition of ERK1 blocks differentiation (Chen et al.)
- Promotes self-renewal and maintains mouse embryonic stem (ES) cells in an undifferentiated, pluripotent state in the absence of feeder cells, serum and leukemia inhibitory factor (LIF) for at least 10 passages (Chen et al.; Xiong et al.)
- When combined with LIF, improves efficiency of ES cell derivation from refractory mouse strains (Pieters et al.; Yang et al.)
Tekniske data
Tekniske data
| CAS | 839707-37-8 |
| Synonym | SC-1 |
| Mængde | 1 mg |
| Molekylær formel | C27H25F3N8O2 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?