Få mere at vide
Novus Biologicals™ Pluripotent Stem Cell Transcription Factor Antibody Pack
Shop alle Bio Techne produkterBeskrivelse
Indeholder
- 1.NB100-2379: OCT4 Antibody
- 2.NB100-588: Nanog Antibody
- 3.NB110-37235: SOX2 Antibody
Pluripotent Stem Cell Transcription Factor Polyclonal antibody specifically detects Pluripotent Stem Cell Transcription Factor in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Tekniske data
Tekniske data
| Klassifikation | Polyclonal |
| Fortynding | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Oprensningsmetode | Affinity Purified |
| Testspecificitet | NB100-58842 is specific for mouse Nanog protein. NB100-2379-OCT4 - Embryonic Stem Cell Marker NB110-37235 - SOX2 - Embryonic Stem Cell Marker |
| Målarter | Mouse, Bovine, Chicken, Primate |
| Indhold og opbevaring | Store at 4C. Do not freeze. |