Learn More
Trajan Scientific and Medical™ Plunger Replacement for Thermo RSH and CTC RTC Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
Brand: Trajan Scientific and Medical™ 0355192
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For sue with Thermo Sceintific instruments
Specifications
PTFE (Plunger) | |
50 μL | |
Gas Tight |
RSH Autosampler Syringe | |
Thermo RSH and CTC RTC Autosampler Syringe |