Learn More
Trajan Scientific and Medical™ Plunger replacement for Shimadzu autosampler syringe PTFE plunger tip 10μL SGE Diamond MS
SGE syringe replacement plungers for gas tight syringes
Brand: Trajan Scientific and Medical™ 0355180
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For use with Shimadzu Autosampler Syringes
Specifications
PTFE (Plunger) | |
10 μL | |
Gas Tight |
Replacement Plunger | |
Shimadzu Autosampler Syringe |