Learn More
Trajan Scientific and Medical™ Plunger Replacement for CTC PAL Autosampler Syringes
SGE syringe replacement plungers for Diamond MS gas tight syringes
Brand:  Trajan Scientific and Medical™ 0355170
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13147966
  • 535.00 DKK / Each
Estimated Shipment: 02-07-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
Specifications
Specifications
PTFE (Plunger)
10 μL
Gas Tight
Replacement Plunger
CTC PAL Autosampler Syringe