Learn More
Trajan Scientific and Medical™ Plunger Replacement for CTC PAL Autosampler Syringes
SGE syringe replacement plungers for Diamond MS gas tight syringes
Brand: Trajan Scientific and Medical™ 0355172
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
Specifications
PTFE (Plunger) | |
50 μL | |
Gas Tight |
Replacement Plunger | |
CTC PAL Autosampler Syringe |