Learn More
Trajan Scientific and Medical™ Plunger Replacement for Agilent Autosampler Syringes
SGE syringe replacement plungers for Diamond MS gas tight syringes.
Brand:  Trajan Scientific and Medical™ 0355160
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13177956
  • 465.00 DKK / Each
Estimated Shipment: 14-06-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For use with Diamond MS Syringes
Specifications
Specifications
Replacement Plunger
10 μL
Agilent Autosampler Syringes