missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLK3, GST Tag Recombinant Protein
PLK3 is a member of the polo family of serine/threonine kinases.
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_004064 |
---|---|
Gene Alias | PLK3, CNK |
KinaseFamily | PLK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408563
|
Invitrogen™
PR7316B |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10587733
|
Invitrogen™
PV4039 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PLK3 is involved in the regulation of M phase in the cell cycle.
Specifications
NP_004064 | |
PLK Kinase Family | |
Recombinant | |
Baculovirus |
PLK3, CNK | |
Human | |
GST Tag |