missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLK1, His Tag Recombinant Protein
PLK1 Recombinant Human Protein
Brand: Invitrogen™ PV3501
Additional Details : Weight : 0.05250kg
Description
PLK1 is a serine⁄threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis.Specifications
NP_005021 | |
5347 | |
10μg | |
PLK Kinase Family | |
Recombinant | |
Storage Buffer: 20 mM Hepes, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.5 mM EGTA, 0.05% Triton X-100, 5 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Other | |
Z'-LYTE™ | |
polo-like kinase 1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
73.3 kDa | |
PLK1 | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
PLK1 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.