Få mere at vide
Tocris Bioscience™ Plitidepsin
Beskrivelse
In vitro, it inhibits induction of ornithine decarboxylase (ODC), which is essential for cell transformation and proliferation.
In vivo, daily administration of Plitidepsin on mice affected by Ehrlich carcinoma duplicated the animal life-span and decreased the total number of tumor cells by 70-90%. Plitidepsin also reduces viral SARS-CoV-2 replication in the lungs (mouse).
It is recommended to prepare stock solutions in DMSO.
Tekniske data
Tekniske data
| CAS | 137219-37-5 |
| Mængde | 2 mg |
| Mål | DNA, RNA and Protein Synthesis Inhibitors |
| Molekylær formel | C57H87N7O15 |
| Renhed | 98% |