Få mere at vide
Plifenat, TRC
Artikelnummer. 30423266
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30423266 1 mg
1 options
Artikelnummer. 30423266 Leverandør Toronto Research Chemicals Leverandørnr. TRCP6320031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Plifenate
CAS 21757-82-4
Molekylær formel C10 H7 Cl5 O2
Mængde 1 mg
Synonym Benzenemethanol','3,4-dichloro-α-(trichloromethyl)-','1-acetate (ACI), Benzenemethanol','3,4-dichloro-α-(trichloromethyl)-','acetate (9CI), Benzyl alcohol','3,4-dichloro-α-(trichloromethyl)-','acetate (8CI), (±)-Acetofenate, 1-(3,4-Dichlorophenyl)-2,2,2-trichloroethyl acetate, 2,2,2-Trichloro-1-(3,4-dichlorophenyl)ethyl acetate, Acetofenate, BAY-MEB 6046, Baygon MEB, MB 6046, Penfenate, Plifenate, β,β,β-Trichloro-α-(3,4-dichlorophenyl)ethanol acetate, β,β,β-Trichloro-α-(3,4-dichlorophenyl)ethyl acetate
InChI formel InChI=1S/C10H7Cl5O2/c1-5(16)17-9(10(13,14)15)6-2-3-7(11)8(12)4-6/h2-4,9H,1H3
SMIL CC(=O)OC(c1ccc(Cl)c(Cl)c1)C(Cl)(Cl)Cl
IUPAC navn [2,2,2-trichloro-1-(3,4-dichlorophenyl)ethyl] acetate
Molekylvægt (g/mol) 336.43
Formel vægt 333.8889