Få mere at vide
Pleconaril-d8 (Major), TRC
Artikelnummer. 30426891
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30426891 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426891 Leverandør Toronto Research Chemicals Leverandørnr. TRCP58035210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pleconaril-d8 (Major)
CAS 1346602-36-5
Molekylær formel C18H10D8F3N3O3
Mængde 10 mg
Synonym 3-[3,5-Dimethyl-4-[3-(3-methyl-5-isoxazolyl)propoxy]phenyl]-5-(trifluoromethyl)-1,2,4-oxadiazole-d8, Picovir-d8, VP 63843-d8, Win 63843-d8
InChI formel InChI=1S/C18H18F3N3O3/c1-10-7-13(16-22-17(27-24-16)18(19,20)21)8-11(2)15(10)25-6-4-5-14-9-12(3)23-26-14/h7-9H,4-6H2,1-3H3/i1D3,2D3,7D,8D
SMIL [2H]c1c(c([2H])c(c(OCCCc2onc(C)c2)c1C([2H])([2H])[2H])C([2H])([2H])[2H])c3noc(n3)C(F)(F)F
Molekylvægt (g/mol) 389.4
Formel vægt 389.1802