Få mere at vide
Platonin Diiodide, TRC
Artikelnummer. 30418514
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418514 100 mg
30422296 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418514 Leverandør Toronto Research Chemicals Leverandørnr. TRCP578210100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3571-88-8
Molekylær formel C38H61I2N3S3
Molekylvægt (g/mol) 909.91
Synonym 4,4'-Dimethyl-3,3'-di-n-heptyl-8-[2-(4-methyl-3-n-heptylthiazole)]2,2'-dicarbocyanin diiodide, 4,4'-Dimethyl-3,3'-dihepthyl-8-[2-(4-methyl-3-heptylthiazole)]-2,2'-dicarbocyanine diiodide, 4,4'-Dimethyl-3,3'-diheptyl-8-[2-(4-methyl-3-heptylthiazole)]-2,2'-dicarbocyanine diiodide, Kankohso 101, NK 19, Photosensitizer 101, Platonin J, Platonin Diiodide, Platonin Iodide, 2,2'-[3-[2-(3-Heptyl-4-methyl-4-thiazolin-2-ylidene)ethylidene]propenylene]bis[3-heptyl-4-methyl-thiazolium Diiodide
IUPAC navn 2-[3,5-bis(3-heptyl-4-methyl-1,3-thiazol-3-ium-2-yl)penta-2,4-dienylidene]-3-heptyl-4-methyl-1,3-thiazole;diiodide
SMIL [I-].[I-].CCCCCCCN1C(=CSC1=CC=C(C=Cc2scc(C)[n+]2CCCCCCC)c3scc(C)[n+]3CCCCCCC)C
Tekniske data
Kemisk navn eller materiale Platonin Diiodide
Mængde 100 mg
InChI formel InChI=1S/C38H61N3S3.2HI/c1-7-10-13-16-19-26-39-32(4)29-42-36(39)24-22-35(38-41(34(6)31-44-38)28-21-18-15-12-9-3)23-25-37-40(33(5)30-43-37)27-20-17-14-11-8-2;;/h22-25,29-31H,7-21,26-28H2,1-6H3;2*1H/q+2;;/p-2
Formel vægt 909.2117