missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on activated carbon paste, Type, Thermo Scientific Chemicals

Catalog Number 15407807
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Physical Form Solid
Quantity 5 g
Odor Odorless
Chemical Name or Material Platinum, 5% on activated carbon paste, Type 128M

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'