Learn More
Platinum, 1% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Product Code. 11394427
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394427 25 g 25g
11304437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11394427 Supplier Thermo Scientific Alfa Aesar Supplier No. 038312.14
 Restricted Item
På lager
Add to basket
CAS: 6-6-7440
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6-6-7440
MDL Number MFCD00011179
Specifications
Quantity 25 g
Chemical Name or Material Platinum, 1% on activated carbon powder, Standard, reduced, nominally 50% water wet