missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 1% on activated carbon paste, Type 163, Thermo Scientific™

Catalog Number 15447697
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Gray
Physical Form Solid
Quantity 5 g
Odor Odorless
Chemical Name or Material Platinum, 1% on activated carbon paste, Type 163
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'