Få mere at vide
MedChemExpress Platelet Factor 4 (58-70), human
Artikelnummer. 16432268
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16432268 5 mg
1 options
Artikelnummer. 16432268 Leverandør MedChemExpress Leverandørnr. HYP1798
MedChemExpress Platelet Factor 4 (58-70), human, a peptide based on the amino acid sequence corresponding to residues 58-70 of platelet factor-4 (PF-4), contains the major heparin-binding domain, which is not sufficient for full antiangiogenic activity.
Tekniske data
CAS 82989-21-7
Indhold og opbevaring Please store the product under the recommended conditions in the Certificate of Analysis.
Til brug med (applikation) Neuroscience-Neuromodulation
Molekylvægt (g/mol) 1572.97 g/mol
Produkttype Peptides