Learn More
Buerkle™ LaboPlast™ Plastic Splinter-Proof Titrating Burettes
Product Code. 11981501 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
15 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11981501 25 mL Each
15591751 15 mL Each
11991501 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11981501 Supplier Buerkle™ Supplier No. 96953025
  • ONLINE EXCLUSIVE PRICE
    625.00 DKK
    595.90 DKK / Pakke med 1
    Spare 29.10 DKK
    4% Slukket
Forventet forsendelse: 23-12-2026
Allows for precise dosing and exact fine titration with the press button or micro-screw and precise discharge through the discharge jet. Buerkle™ Titrating Burets with Shatter Protection has a plastic sheathing which increases resistance to breaki
  • Burette with splinter-proof sheathing
  • Shatter-proof, resistant to chemicals
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
Height (Metric) 65 cm
Includes Storage bottle and Immersion tube, splitter-proof sheathing, Schellbach-stripes
Precision 0.05 mm
Type Titrating Buret
Class Class B
Certifications/Compliance DIN