Beskrivelse
- For various uses
- Acc. to DIN 12331 and ISO 7056
- Blue graduation printed on
- Shatterproof
- Autoclavable
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 100 mL |
| Materiale | Polypropylene |
| Type | Griffin Beaker |
| Autoklaverbar | Autoclavable |
| Engangs | Reusable |
| Uddannet | Yes |
| Skala | Blue Printed Scale |
| Stil | Griffin |