missing translation for 'onlineSavingsMsg'
Learn More

Human PKN1 (PRK1), GST Tag Recombinant Protein

Human PKN1 (PRK1), GST Tag Recombinant Protein

Brand:  Invitrogen PV3790

Additional Details : Weight : 0.05250kg

 View more versions of this product

Product Code. 11513537

  • 4275.00 DKK / 10µg
Estimated Shipment: 18-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PKN1 (PRK1) belongs to the protein kinase C superfamily. This kinase is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids.
Specifications

Specifications

NP_998725
10725, 5585
10 μg
PKN Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: AGC Group
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
protein kinase N1
Kinases & Inhibitors
Liquid
Signal Transduction
Kinase Assay
132.1 kDa
PKN1 (PRK1)
Human
GST Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
PKN1
Baculovirus
Recombinant, Full Length
Serine/Threonine Kinases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers