Få mere at vide
Tocris Bioscience™ PKI (5-24)
High affinity PKA inhibitor
Brand: Tocris Bioscience™ 6221/1
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PKI (5-24) is a high affinity PKA inhibitor (Ki = 2.3 nM).Tekniske data
| Human | |
| 99534-03-9 | |
| 2222.4g/mol | |
| TTYADFIASGRTGRRNAIHD |
| 0.95 | |
| Store at −20°C | |
| 1 mg | |
| High affinity PKA inhibitor (Ki = 2.3nM) |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only