missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC iota Mouse anti-Human, Canine, Feline, Clone: PRKCI/4911, Novus Biologicals™
Mouse Monoclonal Antibody
1940.00 DKK - 3855.00 DKK
Specifications
Antigen | PKC iota |
---|---|
Clone | PRKCI/4911 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310522
|
Bio-Techne
NBP3-13752-20UG |
20 ÎĽg |
1940.00 DKK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18373144
|
Bio-Techne
NBP3-13752-100UG |
100 ÎĽg |
3855.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC iota Monoclonal antibody specifically detects PKC iota in Human, Canine, Feline samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
PKC iota | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Protein Kinase, Signal Transduction | |
10 mM PBS with 0.05% BSA | |
aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota | |
Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743) | |
Primary | |
Store at 4°C. |
PRKCI/4911 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human, Canine, Feline | |
P41743 | |
5584 | |
IgG2b Îş | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PKC iota Mouse anti-Human, Canine, Feline, Clone: PRKCI/4911, Novus Biologicals™