missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ PKA gamma Recombinant Rabbit Monoclonal Antibody (K01_1X10)
GREENER_CHOICE

Artikelnummer. 30827862
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30827862 100 μL
30826596 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30827862 Leverandør Invitrogen™ Leverandørnr. MA563734

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ISINEKCAKE FSEF.

PRKACG (PRKAC gamma) is a third isoform of the catalytic subunit of cAMP-dependent protein kinase. It was isolated from a human testis cDNA library and was clearly derived from a gene distinct from C-alpha and C-beta and showed tissue-specific expression. Whereas at the amino acid level C-alpha and C-beta showed 93% homology, C-gamma showed only about 80% homology to both C-alpha and C-beta. The PRKACG gene is intronless, contains remnants of a poly(A) tail, is flanked by direct repeats, and is co-linear with the PRKACA gene.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PKA gamma
Applikationer Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_1X10
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene PRKACG
Genadgangsnr. P22612
Gene Alias BDPLT19; cAMP-dependent protein kinase catalytic subunit gamma; KAPG; PKA C-gamma; PKACg; PRKACG; protein kinase cAMP-activated catalytic subunit gamma; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase
Gensymboler PRKACG
Værtsarter Rabbit
Immunogen Recombinant protein of human PRKACG (aa 338-351).
Oprensningsmetode Protein A/G
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5568
Målarter Human, Mouse, Rat
Indhold og opbevaring Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.