Få mere at vide
Tocris Bioscience™ PJ 34 hydrochloride
GREENER_CHOICE
Artikelnummer. 18025812
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18025812 10 mg
18054432 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18025812 Leverandør Tocris Bioscience™ Leverandørnr. 3255/10
 Begrænset vare
Forventet forsendelse: 05-08-2026
Læg i indkøbskurv
Læg i indkøbskurv
Potent PARP inhibitor
Potent inhibitor of poly(ADP-ribose) polymerase (PARP) (EC50 = 20 nM). ∽1000-fold more potent than 3-Aminobenzamide (Cat. No. 0788). Protects primary neuronal cells from oxygen-glucose deprivation in vitro and reduces infarct size following focal cerebral ischemia in vivo. Displays protective effects against cisplatin -induced kidney injury. Also displays activity at Pim-1 and Pim-2 kinases at higher concentrations (IC50 values are 3.7 and 16 μM respectively).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 344458-15-7
Molekylær formel C17H18ClN3O2
Tekniske data
Kemisk navn eller materiale PJ 34 hydrochloride
Mængde 10 mg
Formel vægt 331.8
Renhed >98%
Molekylvægt (g/mol) 331.8