Få mere at vide
Pizotifen Malate, TRC
Artikelnummer. 30413385
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30413385 10 mg
1 options
Artikelnummer. 30413385 Leverandør Toronto Research Chemicals Leverandørnr. TRCP55280310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pizotifen Malate
CAS 5189-11-7
Molekylær formel C19 H21 N S . C4 H6 O5
Mængde 10 mg
Synonym Butanedioic acid','2-hydroxy-','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1), Butanedioic acid','hydroxy-','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1) (9CI), Malic acid','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1) (8CI), Malic acid','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (7CI), 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene','9,10-dihydro-4-(1-methyl-4-piperidylidene)-','malate (1:1) (8CI), 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene','piperidine deriv., Piperidine','4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-','hydroxybutanedioate (1:1) (9CI), Piperidine','4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-','malate (1:1) (8CI), 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidinium hydrogen malate, Mosegor, Pizotifen hydrogen malate, Pizotifen malate, Pizotyline malate, Sanmigran, Sanomigran
InChI formel InChI=1S/C19H21NS.C4H6O5/c1-20-11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-21-18;5-2(4(8)9)1-3(6)7/h2-5,10,13H,6-9,11-12H2,1H3;2,5H,1H2,(H,6,7)(H,8,9)
SMIL CN1CCC(=C2c3ccccc3CCc4sccc24)CC1.OC(CC(=O)O)C(=O)O
IUPAC navn 4-(4,5-dihydrobenzo[1,2]cyclohepta[2,4-b]thiophen-10-ylidene)-1-methylpiperidine;2-hydroxybutanedioic acid
Molekylvægt (g/mol) 429.53
Formel vægt 429.161