Få mere at vide
Pivalic acid-d9, TRC
Artikelnummer. 30422721
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422721 250 mg
30417157 500 mg
30410101 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422721 Leverandør Toronto Research Chemicals Leverandørnr. TRCP520004250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42983-07-3
Molekylær formel C5 D9 H O2
Molekylvægt (g/mol) 111.19
Synonym 2,2-Di(methyl-d3)propanoic-3,3,3-d3 Acid, 2,2-Dimethylpropanoic Acid-d9, 2,2,2-Trimethylacetic Acid-d9, 2,2-Dimethylpropanoic Acid-d9, 2,2-Dimethylpropionic Acid-d9, NSC 65449-d9, Neopentanoic Acid-d9, Neovaleric Acid-d9, Trimethylacetic Acid-d9, Trimethylmethanecarboxylic Acid-d9, Versatic 5 Acid-d9, tert-Pentanoic Acid-d9, alpha,alpha-Dimethylpropionic Acid-d9, Trimethyl-d9-acetic Acid
IUPAC navn 3,3,3-trideuterio-2,2-bis(trideuteriomethyl)propanoic acid
SMIL [2H]C([2H])([2H])C(C(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Pivalic acid-d9
Mængde 250 mg
InChI formel InChI=1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)/i1D3,2D3,3D3
Formel vægt 111.1246