Learn More
PITPNC1 Antibody, PerCP, R&D Systems™
Product Code. 18604622 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. Unit Size
18604622 0.1mL
1 options
Product Code. 18604622 Supplier Novus Biologicals Supplier No. NBP297834PCP
 Restricted Item
Forventet forsendelse: 11-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
PITPNC1 Polyclonal specifically detects PITPNC1 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin)/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Product Type Antibody
Product Line Novus