Få mere at vide
Pitavastatin Calcium, TRC
Artikelnummer. 30425958
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425958 50 mg
30409131 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30425958 Leverandør Toronto Research Chemicals Leverandørnr. TRCP53100050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pitavastatin Calcium
CAS 147526-32-7
Renhedsgrad noter HPLC
Molekylær formel 2 C25 H23 F N O4 . Ca
Mængde 50 mg
Synonym (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Calcium Salt (2:1), 6-Heptenoic acid','7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-','calcium salt (2:1)','[S-[R*,S*-(E)]]-, (E)-(3R,5S)-7-[2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoic acid hemicalcium salt, Flovas, Livalo, NK 104, NK 104 (acid), Pitava, Pitava 1, Pitavastatin calcium, Pitavastatin hemicalcium
InChI formel InChI=1S/2C25H24FNO4.Ca/c2*26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31;/h2*1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31);/q;;+2/p-2/b2*12-11+;/t2*18-,19-;/m11./s1
SMIL [Ca+2].O[C@H](C[C@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4)CC(=O)[O-].O[C@H](C[C@H](O)\C=C\c5c(nc6ccccc6c5c7ccc(F)cc7)C8CC8)CC(=O)[O-]
IUPAC navn calcium;(E,3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate
Molekylvægt (g/mol) 880.98
Formel vægt 880.2848
Procent renhed >95