Få mere at vide
Piroxicam EP Impurity D, Mikromol™
Artikelnummer. 17991516
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991516 25 mg
17964366 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991516 Leverandør Mikromol Leverandørnr. MM0084.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Piroxicam EP Impurity D
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
CAS 6639-62-9
Analyt- eller komponentnavne Methyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
Forsendelsestilstand Room Temperature
Molekylær formel C10 H9 N O5 S
Certificeringer/compliance ISO 9001
Synonym Methyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Piroxicam Imp. D (EP), Piroxicam Impurity D
InChI formel InChI=1S/C10H9NO5S/c1-16-9(12)6-11-10(13)7-4-2-3-5-8(7)17(11,14)15/h2-5H,6H2,1H3
SMIL COC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC navn methyl 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetate
Molekylvægt (g/mol) 255.25
Mængde 25 mg
Formel vægt 255.0201 g/mol