Få mere at vide
Piroxicam-d3, TRC
Artikelnummer. 30412846
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412846 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412846 Leverandør Toronto Research Chemicals Leverandørnr. TRCP5100021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piroxicam-d3
CAS 942047-64-5
Renhedsgrad noter HPLC
Molekylær formel C15 2H3 H10 N3 O4 S
Mængde 1 mg
Synonym 4-Hydroxy-2-methyl-d3-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, Piroxicam-d3
InChI formel InChI=1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)/i1D3
SMIL [2H]C([2H])([2H])N1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ccccn3
IUPAC navn 4-hydroxy-1,1-dioxo-N-pyridin-2-yl-2-(trideuteriomethyl)-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvægt (g/mol) 334.36
Formel vægt 334.0815
Procent renhed >95