missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Pirfenidone, MedChemExpress
Beskrivelse
Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pirfenidone |
| Fysisk form | Powder |
| CAS | 53179-13-8 |
| Farve | White |
| Mængde | 5 g |
| Renhedsgrad noter | Research |
| Molekylær formel | C12H11NO |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | AMR69 |
| Opløselighedsinformation | DMSO : ≥ 100 mg/mL (539.90 mM) ∣H2O : 16.67 mg/mL (90.00 mM; Need ultrasonic) |
| Vis mere |
Research purposes only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?