Få mere at vide
Piretanide-d4 (Major), TRC
Artikelnummer. 30425547
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30425547 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425547 Leverandør Toronto Research Chemicals Leverandørnr. TRCP5087021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Piretanide-d4 (Major)
CAS 1246816-90-9
Molekylær formel C17H14D4N2O5S
Mængde 1 mg
Synonym 3-(Aminosulfonyl)-4-phenoxy-5-(1-pyrrolidinyl-d4)benzoic Acid, Arelix-d4, Arlix-d4, Diumax-d4, Eurelix-d4, HOE 118-d4, S 73-4118-d4, Tauliz-d4
InChI formel InChI=1S/C17H18N2O5S/c18-25(22,23)15-11-12(17(20)21)10-14(19-8-4-5-9-19)16(15)24-13-6-2-1-3-7-13/h1-3,6-7,10-11H,4-5,8-9H2,(H,20,21)(H2,18,22,23)/i8D2,9D2
SMIL [2H]C1([2H])CCC([2H])([2H])N1c2cc(cc(c2Oc3ccccc3)S(=O)(=O)N)C(=O)O
IUPAC navn 4-phenoxy-3-sulfamoyl-5-(2,2,5,5-tetradeuteriopyrrolidin-1-yl)benzoic acid
Molekylvægt (g/mol) 366.42
Formel vægt 366.1187